Learn More
Tocris Bioscience™ Endothelin 2 (human)
GREENER_CHOICE
Catalog No. 1164100U
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
1164100U 100 μg
1 options
This item is not returnable. View return policy
Catalog No. 1164100U
Supplier: Tocris Bioscience™ 1164/100U
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
ETA/ETB agonist
Endothelin 2 (human) is an endogenous peptide found predominately in the kidney and intestine. Displays similar selectivity for ETA and ETB endothelin receptors.