Learn More
        Tocris Bioscience™ Galanin (1-29) (rat, mouse)
                    Click to view available options
                                Quantity:
                                1 mg
                    Description
Galanin (1-29) (rat, mouse) is a non-selective galanin receptor agonist (Ki values are 0.98, 1.48 and 1.47 nM for GAL1, GAL2 and GAL3 respectively). Anticonvulsant; prevents the occurrence of full kindled seizures in rats.
            Specifications
Specifications
| Purity | 95% | 
| Quantity | 1 mg | 