Learn More
Tocris Bioscience™ [Leu31,Pro34]-Neuropeptide Y (porcine)
Catalog No. 1168200U
Click to view available options
Quantity:
200 μg
This item is not returnable. View return policy
This item is not returnable. View return policy
NPY Y1 receptor agonist
High affinity neuropeptide Y Y1 receptor agonist (Ki = 0.54 nM). Also shows affinity for Y5 receptors.
Specifications
Quantity 200 μg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.