Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ M40

Click to view available options
Quantity:
1 mg
Description
M40 is a potent, non-selective galanin receptor antagonist (Ki values are 1.82 and 5.1 nM at GAL1 and GAL2 respectively) that inhibits galanin (1-29) (Cat. No. 2696) binding in rat brain in vitro (IC50 = 3 - 15 nM). Attenuates the antidepressant effects of fluoxetine (Cat. No. 0927) and blocks galanin-induced food intake in vivo. Also exhibits weak partial agonist activity at peripheral GAL2 receptors at doses > 100 nM.
Specifications
Specifications
Purity | 0.95 |
Content And Storage | Store at -20°C |
Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction