Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Neuromedin B (porcine)
Click to view available options
Quantity:
1 mg
Description
Neuromedin B (porcine) is a mammalian bombesin-like peptide and endogenous ligand for the neuromedin B receptor (IC50 = 2 nM); first isolated from pig spinal cord. Regulates endocrine and exocrine secretion, smooth muscle contraction, blood pressure, feeding and cell growth.
Specifications
Specifications
Purity | 95% |
Content And Storage | Store at -20°C |
Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction