Learn More
Tocris Bioscience™ Peptide YY (3-36)
GSA_VAGREENER_CHOICE
Catalog No. 1618500U
Click to view available options
Quantity:
500 μg
This item is not returnable. View return policy
This item is not returnable. View return policy
Selective NPY Y2 receptor agonist
Peptide YY (3-36) is a Y2 selective agonist. IC50 values are 0.11 and 1050 nM for inhibition of 125I-PYY binding to Y2 and Y1 receptors respectively. Inhibits food intake and reduces weight gain in vivo. Brain penetrant.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 500 μg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.