Learn More
Tocris Bioscience™ PKA inhibitor fragment (6-22) amide
GREENER_CHOICE
Catalog No. 19041
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy
This item is not returnable. View return policy
Potent protein kinase A inhibitor
PKA inhibitor fragment (6-22) amide is a potent inhibitor of cAMP-dependent protein kinase (PKA) (Ki = 2.5 nM); derived from the active portion of the heat-stable PKA inhibitor protein PKI.
Specifications
Quantity 1 mg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.