Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Urocortin (human)
GREENER_CHOICE

Catalog No. 16041
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Endogenous CRF agonist

Urocortin (human) is an endogenous CRF agonist. Ki values are 0.4, 0.3 and 0.5 nM for hCRF1, rCRF2α and mCRF2β respectively.

Specifications

Purity 95%
Content And Storage Store at -20°C
Quantity 1 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.