Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Todd Hewitt Broth with Gentamicin and Nalidixic Acid (Trans Vag Broth)

Catalog No. r064840
Click to view available options
Quantity:
100/Pk.
20/Pk.
Format:
Tube
Media Type:
Trans Vag Broth
Includes:
Gentamicin and Nalidixic Acid
This item is not returnable. View return policy
This item is not returnable. View return policy

Reduce media prep time while selectively isolating group B streptococci from clinical specimens containing mixed flora with pre-filled, ready-to-use tubes.

Selectively isolate group B streptococci from clinical specimens containing mixed flora using Thermo Scientific™ Todd Hewitt Broth with Gentamicin and Nalidixic Acid (Trans Vag Broth).

Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes of Todd Hewitt Broth with the addition of Gentamicin and Nalidixic Acid to inhibit growth of some gram-negative bacteria. Requires the addition of 0.25mL of 5% sheep blood per tube before use to increase recovery of group B streptococci.

Streptococci are facultatively anaerobic, catalase-negative, gram-positive cocci that occur as pairs or chains. They cause a wide variety of clinical diseases including pharyngitis, scarlet fever, otitis media, impetigo, meningitis, neonatal sepis, pneumonia, endocarditis, osteomyelitis, and necrotizing fascitis. One of the most useful phenotypic characteristics of streptococci is the hemolytic reaction, generally classified as alpha, beta or gamma according to the appearance of zones around colonies growing on Blood Agar. Beta-hemolytic streptococci are further characterized by Lancefield groups based on the antigenic differences in group-specific polysaccharides located in the bacterial cell wall.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear liquid, light amber
Content And Storage 2°C to 8°C
Format Tube
Manufacturing Origin United States
Media Type Trans Vag Broth
Product Type Microbiology Media
Fill Volume 5 mL
Includes Gentamicin and Nalidixic Acid
Packaging Type Glass tube with red screw cap
Quantity 100/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.