Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564401
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564401 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564401 Supplier Invitrogen™ Supplier No. PA564401
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LHRAGFDAFM TGYVMAYVEV SQGPQPCSSG PWLPECHNKV YLSGKAVPLT VAKSQFSRSS KAHNQKMKLT WGS Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

Inhibits cell growth rate and cell cycle. Induces CDKN1A expression as well as TGF-beta expression. Mediates the inhibitory growth effect of EGR1. Involved in the maturation of snRNAs and snRNA 3'-tail processing (PubMed:28092684). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOE1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TOE1
Gene Accession No. Q96GM8
Gene Alias 4930584N22Rik; 4933424D16Rik; AI413517; hCaf1z; Target of EGR1 protein 1; target of EGR1, member 1 (nuclear); TOE1
Gene Symbols TOE1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TOE1. Recombinant protein control fragment (Product #RP-103969).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114034
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.