Description
Impacto™ Toes2Go steel toe cap overshoes
- Made of flexible waterproof that do not harden or crack
- Treads under sole help prevent slipping
- For visitors, casual or temporary workers
- Fits tightly over existing shoes, providing a solid protecting cap
- Comfortably worn over low heel, dress, running shoes and most boots
- Color-coded for sizing
- Meet ASTM F2412/F2413 impact and compression standard
- Fully Adjustable 18 x 1 inch strap with hook and loop closure
Specifications
Specifications
| Color | Red |
| Material | PVC |
| Size | Men: 8 to 13, Women: 10 to 13 |
| Toe Material | Steel |
| Gender | Unisex |
| Waterproof | Yes |
| Product Type | Steel toe cap |