Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BCCIP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565200
Description
Immunogen sequence: ALPMYQQLQKE LAGAHRTNKP CGKCYFYLLI SKTFVEAGKN NSKKKPSNKK KAALMFANAE EEFFYEEQGK PEVLGGPDTR RGLEPVPIQH NGGSR Highest antigen sequence identity to the following orthologs - mouse 57%, rat 54%.
This gene product was isolated on the basis of its interaction with BRCA2 and p21 proteins. It is an evolutionarily conserved nuclear protein with multiple interacting domains. The N-terminal half shares moderate homology with regions of calmodulin and M-calpain, suggesting that it may also bind calcium. Functional studies indicate that this protein may be an important cofactor for BRCA2 in tumor suppression, and a modulator of CDK2 kinase activity via p21. This protein has also been implicated in the regulation of BRCA2 and RAD51 nuclear focus formation, double-strand break-induced homologous recombination, and cell cycle progression. Multiple transcript variants encoding different isoforms have been described for this gene.
Specifications
BCCIP | |
Polyclonal | |
Unconjugated | |
BCCIP | |
1110013J05Rik; Bccip; BCCIPalpha; BCCIPbeta; BRCA2 and CDKN1A interacting protein; BRCA2 and CDKN1A-interacting protein; BRCA2 and Cip1/p21 interacting protein; cdk inhibitor p21 binding protein; p21- and CDK-associated protein 1; Protein TOK-1; TOK1; TOK-1; TOK-1alpha; TOK-1beta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56647 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P287 | |
BCCIP | |
Recombinant Human TOK-1. Recombinant protein control fragment (Product #RP-105943). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction