Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Tolcapone
GREENER_CHOICE

Catalog No. 586410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
586410 10 mg
586450 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 586410 Supplier Tocris Bioscience™ Supplier No. 5864/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

COMT inhibitor; also inhibits transthyretin aggregation

Tolcapone is a COMT inhibitor. Inhibits both brain and peripheral COMT. Also binds transthyretin (TTR) with high affinity (Kd1 and Kd2 values are 21 and 58 nM, respectively). Inhibits TTR aggregation in human plasma and prevents TTR-induced cytotoxicity in vitro. Stabilizes TTR in mice and humans in vivo. Orally bioavailable.

Specifications

Chemical Name or Material (3,4-Dihydroxy-5-nitrophenyl)(4-methylphenyl)methanone
CAS 134308-13-7
Synonym Tasmar, Ro 40-7592
Quantity 10 mg
Target Catechol O-Methyltransferase (COMT) Inhibitors
Molecular Formula C14H11NO5
Formula Weight 273.24
Purity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.