Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tolfenamic Acid, 99+%, PROMO, Thermo Scientific Chemicals

Catalog No. J6125609PM
Click to view available options
Quantity:
10 g

NSAID that activates calcium-activated potassium channels

Applications

Tolfenamic acid is a non-steroidal anti-inflammatory agent. It interferes with synthesis of B-amyloid precursor protein, and thus AB peptides, by promoting degradation of an essential transcription factor. It inhibits fMLP- and A23187-induced Ca2+ influx in human PMNL with an IC50 value of approximately 20 μM.

Solubility

Soluble in water (slightly), acetone (approx.10 mg/mL), DMSO (52 mg/mL at 25°C), methanol (approx.10 mg/mL) and ethanol (50 mg/mL).

Notes

Stable under recommended storage conditions. Incompatible with oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13710-19-5
Molecular Formula C14H12ClNO2
Molecular Weight (g/mol) 261.71 g/mol
MDL Number MFCD00133865
InChI Key YEZNLOUZAIOMLT-UHFFFAOYSA-N
Synonym tolfenamic acid, clotam, n-3-chloro-2-methylphenyl anthranilic acid, 2-3-chloro-2-methylphenyl amino benzoic acid, tolfedine, acido tolfenamico, acide tolfenamique, acidum tolfenamicum, tolfine, n-2-methyl-3-chlorophenyl anthranilic acid
PubChem CID 610479
ChEBI CHEBI:32243
IUPAC Name 2-(3-chloro-2-methylanilino)benzoic acid
SMILES CC1=C(C=CC=C1Cl)NC2=CC=CC=C2C(=O)O

Specifications

Color White
Quantity 10 g
UN Number UN2811
Merck Index 14,9513
Solubility Information Soluble in water, acetone, DMSO, methanol and ethanol.
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material Tolfenamic acid
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 237-264-3
RTECSNumber CB2687500
TSCA No

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.