Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tollip Antibody [DyLight 350], Novus Biologicals™

Sheep Polyclonal Antibody
Supplier: Novus Biologicals AF4678D
Description
Tollip Polyclonal antibody specifically detects Tollip in Human samples. It is validated for Western BlotSpecifications
Tollip | |
Polyclonal | |
50mM Sodium Borate | |
Sheep | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 350 | |
adapter protein, FLJ33531, IL-1RAcPIP, toll interacting protein, toll-interacting protein | |
E. coli-derived recombinant human Tollip, Met1-Pro274, Accession # Q9H0E2 | |
0.1 mL | |
Signal Transduction | |
54472 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction