Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tollip Antibody (Kimmy-2), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197632
Description
Tollip Monoclonal specifically detects Tollip in Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
Tollip | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1 ug/ml, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500 | |
TOLLIP | |
Recombinant mouse Tollip (aa 1-274). | |
0.05 mg | |
Primary | |
Mouse. There is a weak cross-reactivity to Human Tollip. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Kimmy-2 | |
Unconjugated | |
adapter protein, FLJ33531, IL-1RAcPIP, toll interacting protein, toll-interacting protein | |
Mouse | |
Tissue culture supernatant | |
RUO | |
54472 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction