Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tollip Antibody (SB40a), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP128621PEATT594
Description
Tollip Monoclonal specifically detects Tollip in Human samples. It is validated for Flow Cytometry.Specifications
Tollip | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
TOLLIP | |
Recombinant Tollip | |
0.1 mL | |
Primary | |
Human Tollip; Does not react to TLR2, TLR4, TLR6, TLR7, TLR8 and TLR10 recombinant proteins | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
SB40a | |
Flow Cytometry | |
adapter protein, FLJ33531, IL-1RAcPIP, toll interacting protein, toll-interacting protein | |
Mouse | |
Protein A or G purified | |
Signal Transduction | |
54472.0 | |
Human | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction