Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tolnaftate, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18607446 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
1 g
10 g
50 g
250 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail

TO102, 2398-96-1, C19H17NOS

Spectrum™ Chemical Tolnaftate, USP is an antifungal medication that fights infections caused by fungus such as athlete's foot and jock itch. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 2398-96-1
Molecular Formula C19H17NOS
Molecular Weight (g/mol) 307.41
InChI Key FUSNMLFNXJSCDI-UHFFFAOYSA-N
IUPAC Name N-methyl-N-(3-methylphenyl)-1-(naphthalen-2-yloxy)methanethioamide
SMILES CN(C(=S)OC1=CC=C2C=CC=CC2=C1)C1=CC=CC(C)=C1

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Melting Range 110-113°C
Packaging Amber Glass Bottle
Quantity 1 g
Percent Purity 98 to 102%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.