Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tolnaftate, USP, 98-102%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
1 g
10 g
50 g
250 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
Description
Spectrum™ Chemical Tolnaftate, USP is an antifungal medication that fights infections caused by fungus such as athlete's foot and jock itch. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 1 |
| Ignition Residue | 0.001 |
| Loss on Drying | 0.005 |
| Melting Range | 110-113°C |
| Packaging | Poly Pail |
| Quantity | 5 kg |
| Percent Purity | 98 to 102% |
| Grade | USP |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction