Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium carbonate, 99.99%, (trace metal basis)

Catalog No. AC278020500
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle

For ultra trace analysis | CAS: 534-17-8 | CCs2O3 | 325.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-17-8
Molecular Formula CCs2O3
Molecular Weight (g/mol) 325.82
MDL Number MFCD00010957
InChI Key FJDQFPXHSGXQBY-UHFFFAOYSA-L
Synonym cesium carbonate, dicesium carbonate, caesium carbonate, cesiumcarbonate, carbonic acid, dicesium salt, cs2co3, unii-qqi20a14p4, cesium carbonate cs2co3, carbonic acid, cesium salt 1:2, dicaesium 1+ ion carbonate
PubChem CID 10796
SMILES [Cs+].[Cs+].[O-]C([O-])=O

Specifications

Melting Point >400.0°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.99%
Linear Formula Cs2CO3
Fieser 07,57; 08,81; 09,99; 12,108; 13,70; 14,77; 15,73
Merck Index 15, 2009
Solubility Information Solubility in water: 261 g/100mL water (20°C). Other solubilities: soluble in alcohol and ether
IUPAC Name dicesium;carbonate
Formula Weight 325.82
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 4.072
Density 4.0720g/mL
Chemical Name or Material Cesium carbonate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Suspected of damaging fertility or the unborn child.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure if swallowed.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
EINECSNumber 208-591-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.