Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, 99+%, extra pure

Catalog No. p-3523604
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
25 L
Packaging:
Glass Bottle
Metal Drum
Plastic Drum

CAS: 108-88-3 | C7H8 | 92.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -95.0°C
Density 0.8660g/mL
Boiling Point 111.0°C
Quantity 1 L
Flash Point 4°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4968
Residue after Evaporation 0.003% max.
Assay Percent Range 99+%
Specific Gravity 0.866
Linear Formula C6H5CH3
Vapor Pressure Vapor Pressure: 29mbar at 20°C
Viscosity 0.6 mPa.s (20°C)
Beilstein 05, 280
Water 0.05% max. (K.F.)
Merck Index 15, 9688
Solubility Information Solubility in water: almost insoluble (0.5g/L at 20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, acetone,, ether, petroleum ether, benzene, ch2cl2, dmf, gla, cial acetic acid, chcl3, ccl4.
Formula Weight 92.14
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material Toluene
Show More Show Less
Hazard Category Signal Word: DANGER!
Hazard Statement GHS P Statement May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. May be fatal if swallowed and enters airways. Causes skin irritation. Suspected of damag
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.