Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, ACS, 99.5% min

Catalog No. p-4408297
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L

CAS: 108-88-3 | C7H8 | 92.14 g/mol

Toluene is used as solvent for paints, paint thinners, silicone sealants, many chemical reactants, rubber, printing ink, adhesives (glues), lacquers, leather tanners and disinfectants. It is an important raw material to synthesize pharmaceuticals, paints, resins, dyes, toluene diisocyanate and explosives like trinitrotoluene. It is used as standard material for chromatography and analytical reagent. It plays an important role in the manufacture of benzene, xylene, phenol, poly urethane polymers, cosmetics and jet fuel blends. It is used as fuel additive to increase octane ratings and a coolant for its good heat transfer capabilities in sodium cold traps used in nuclear reactor system loops.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point °C to 93°C
Color Colorless
Appearance Colorless liquid
Density 0.867 g/cm3 at 20°C
Boiling Point 111°C
Quantity 500 mL
Flash Point 4°C (40°F)
Odor Aromatic
Physical Form Liquid
Refractive Index 1.4967
Assay Percent Range 99.5%
Vapor Pressure 29 hPa (22mm Hg) at 20°C
Beilstein 635760
Solubility Information Very slightly soluble in water. Miscible with alcohol,chloroform,ether,glacial acetic acid,carbon disulfideMiscible with ethanol,benzene,diethyl ether,acetone,chloroform,glacial acetic acid and carbon disulfide. Immiscible with water.
Formula Weight 92.14
Grade ACS
Chemical Name or Material Toluene
Show More Show Less
Hazard Category H225-H304-H315-H319-H336-H361d-H373
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation DOT Class: 3; Packing Group: II
EINECSNumber 203-625-9
RTECSNumber XS5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.