Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Toluene ACS AR, Macron Fine Chemicals™
Click to view available options
Quantity:
4 L
Packaging:
Glass Container
SAFEMOR
Specifications
Specifications
Quantity | 4 L |
Packaging | Glass Container |
Formula Weight | 92.14 |
Grade | ACS Reagent |
Chemical Name or Material | Toluene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction