Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, anhydrous, 99.8%

Catalog No. aa41464ak
Click to view available options
Quantity:
250 mL
1 L
4 L

CAS: 108-88-3 | C7H8 | 92.14 g/mol

Toluene is widely used as an organic solvent in various industries like paints, pharmaceuticals, dyes, agrochemicals and polymers. It is used as an additive in gasoline and improves octane ratings, as a fullerene indicator, in paint thinners and silicone sealants, as a coolant in sodium cold traps used in nuclear reactor system loops and as a fuel for both two-stroke and four-stroke engines. It is also used as a raw material in the production of benzene, benzaldehyde, benzoic acid, toluene diisocyanate, benzyl-tert-butyl peroxide, xylene and many other chemical products. It plays an important role in biochemistry experiments to break open red blood cells in order to extract haemoglobin as well as used in the removal of cocaine from coca leaves to make coca-cola syrup.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point °C to 93°C
Density 0.867 g/mL
Boiling Point 111°C
Quantity 250 mL
Flash Point 4°C (40°F)
Physical Form Liquid
Refractive Index 1.4967
Assay Percent Range 99.8%
Beilstein 635760
Solubility Information Miscible with ethanol,alcohol,benzene,diethyl ether,acetone,chloroform,glacial acetic acid and carbon disulfide. Immiscible with water.
Formula Weight 92.14
Chemical Name or Material Toluene, anhydrous
Show More Show Less
Hazard Category H225-H304-H315-H319-H336-H361d-H373
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 203-625-9
RTECSNumber XS5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.