Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Toluene, for HPLC, Thermo Scientific Chemicals
Click to view available options
Quantity:
4 L
Packaging:
EcoSafPak™
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC

Specifications
Specifications
Absorbance | (1 cm cell vs water), 1 max. at 286nm |
Density | 0.8660g/mL |
Quantity | 4 L |
Identification | Pass Test |
Packaging | EcoSafPak™ |
Refractive Index | 1.4930 to 1.4950 |
Residue after Evaporation | 10ppm max. |
Linear Formula | C6H5CH3 |
Vapor Pressure | Vapor Pressure: 29mbar at 20°C |
Beilstein | 05, 280 |
Show More |
Safety and Handling
Hazard Category | Signal Word: DANGER! |
Hazard Statement | GHS H Statement: May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure. May be fatal if swallowed and enters airways. Causes skin irritation. Suspected of damaging the unborn child. Highly flammable liquid and vapor. |
Precautionary Statement | Emergency Overview Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects. NFPA Health:2 Flammability:3 Instability:0 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction