Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Toluene, Laboratory Reagent, ≥99.3%, Honeywell™
Click to view available options
Quantity:
1 L
2.5 L
10 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
Description
- ≥95% for general laboratory use.

Specifications
Specifications
Melting Point | -95°C |
Color | Colorless |
Density | 0.865g/mL at 25°C |
Boiling Point | 110.6°C |
Quantity | 2.5 L |
Explosion Limit | 0.07 |
Flash Point | 4°C (39.2°F) |
Ignition Point | 997°F |
Packaging | Glass Bottle |
Physical Form | Liquid |
Show More |
Safety and Handling
missing translation for 'healthHazard2' | P210-P260-P280-P301 + P310-P370 + P378-P403 + P235 |
missing translation for 'healthHazard3' | H225-H304-H315-H336-H361d-H373 |
DOTInformation | class 3 - PG 2 - Toluene |
EINECSNumber | 203-625-9 |
RTECSNumber | XS5250000 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction