Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, Laboratory Reagent, ≥99.3%, Honeywell™

Catalog No. p-7120882
Click to view available options
Quantity:
1 L
2.5 L
10 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can

Laboratory Reagent,≥99.3%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -95°C
Color Colorless
Density 0.865g/mL at 25°C
Boiling Point 110.6°C
Quantity 1 L
Explosion Limit 0.07
Flash Point 4°C (39.2°F)
Ignition Point 997°F
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n/D 1.496(lit.)
Linear Formula C6H5CH3
Vapor Pressure 22mmHg (20°C), 26mmHg (25°C)
UN Number UN1294
Beilstein 635760
Formula Weight 92.14g/mol
Percent Purity ≥99.3%
Grade Laboratory Reagent
Chemical Name or Material Toluene
Show More Show Less
Hazard Statement P210-P260-P280-P301 + P310-P370 + P378-P403 + P235
Precautionary Statement H225-H304-H315-H336-H361d-H373
DOTInformation class 3 - PG 2 - Toluene
EINECSNumber 203-625-9
RTECSNumber XS5250000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.