Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, Technical, Spectrum™ Chemical
SDP

Catalog No. p-7166647 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
55 gal.
Packaging:
Amber Glass Bottle
Drum
Steel Can
Catalog No. Quantity Packaging
18-614-487 500 mL Amber Glass Bottle
18-614-486 4 L Amber Glass Bottle
18-614-485 20 L Steel Can
18-614-488 55 gal. Drum
4 options

Catalog No. 18-614-487

Supplier: Spectrum Chemical Manufacturing Corporation T1092500MLGL

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 108-88-3

Specifications

CAS Max % 1
Boiling Point 111°C
Quantity 500 mL
Packaging Amber Glass Bottle
Grade Technical
Chemical Name or Material Toluene
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Not for pesticide use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.