Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Toluidine Blue O Solution, 0.5%
SCI_ED

Catalog No. s25612 Shop All Fisherbrand Products
Click to view available options
Quantity:
15 mL
100 mL
Packaging:
0.5%

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 92-31-9, 7732-18-5
Molecular Formula C15H16ClN3S
Molecular Weight (g/mol) 305.82
MDL Number MFCD00011934
InChI Key HNONEKILPDHFOL-UHFFFAOYSA-M
PubChem CID 7083
IUPAC Name 3-amino-7-(dimethylamino)-2-methyl-5λ⁴-phenothiazin-5-ylium chloride
SMILES [Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1

Specifications

Assay Percent Range 0.5% w/v
Grade Laboratory
Packaging 0.5%
Density ∽1g/mL
Color Blue/Purple
Quantity 15 mL
Chemical Name or Material Toluidine Blue O Solution, 0.5%
ShelfLife 18 months
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.