Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Tolvaptan
GREENER_CHOICE

Catalog No. 518110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-811-0 10 mg
51-815-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 51-811-0 Supplier Tocris Bioscience™ Supplier No. 5181/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective vasopressin V2 receptor antagonist; renoprotective and orally active

Tolvaptan is a potent and selective competitive vasopressin V2 receptor antagonist (Ki values are 0.06 and 12.3 nM for V2 and V1a receptors respectively). Decreases urine osmolality and increases serum sodium concentrations. Delays the onset of end-stage renal disease in a mouse model of polycystic kidney disease. Exhibits myocardial and renal protective effects in hypertensive heart failure rats. Orally active.

Specifications

Quantity 10 mg
Product Type Tolvaptan
Purity >98%
Molecular Weight (g/mol) 448.94
Inhibitors Tolvaptan

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.