Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tolylene 2,4-diisocyanate, 75%, technical
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 12.5°C to 13.5°C |
| Color | Colorless to Yellow |
| Density | 1.2200g/mL |
| Boiling Point | 115.0°C to 120.0°C (10.0 mmHg) |
| Flash Point | 127°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 75% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.5670 to 1.5690 |
| Linear Formula | CH3C6H3(NCO)2 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes skin irritation. Causes serious eye irritation. Fatal if inhaled. May cause an allergic skin reaction. Suspected of cau |
| Precautionary Statement | GHS P Statement Immediately call a POISON CENTER or doctor/physician. If skin irritation occurs: Get medical advice/attention. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. If skin irritat |
| EINECSNumber | 209-544-5 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction