Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOM1 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10061601AF594
Description
TOM1 Polyclonal antibody specifically detects TOM1 in Human samples. It is validated for Western BlotSpecifications
TOM1 | |
Polyclonal | |
Western Blot | |
ADP-ribosylation factor-binding protein GGA3, golgi associated, gamma adaptin ear containing, ARF binding protein 3, golgi-associated, gamma adaptin ear containing, ARF binding protein 3, KIAA0154Golgi-localized, gamma ear-containing, ARF-binding protein 3 | |
The immunogen recognized by this antibody maps to a region between residue 400 and 450 of human target of myb1 using the numbering given in entry NP_005479.1 (GeneID 10043). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
10043 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction