Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TOM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546711
Description
Peptide sequence: VTSEEFDKFL EERAKAADRL PNLSSPSAEG PPGPPSGPAP RKKTQEKDDD.
This gene was identified as a target of the v-myb oncogene. The encoded protein shares its N-terminal domain in common with proteins associated with vesicular trafficking at the endosome. It is recruited to the endosomes by its interaction with endofin. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
TOM1 | |
Polyclonal | |
Unconjugated | |
TOM1 | |
FLJ33404; target of myb 1; target of Myb protein 1; target of myb1; target of myb1 (chicken); target of myb1 homolog; target of myb1 homolog (chicken); target of myb1 membrane trafficking protein; TOM1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10043 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O60784 | |
TOM1 | |
Synthetic peptide directed towards the C-terminal of human TOM1 (aa 438-487). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction