Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598207
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598207 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598207 Supplier Invitrogen™ Supplier No. PA598207
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TOM1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

This gene was identified as a target of the v-myb oncogene. The encoded protein shares its N-terminal domain in common with proteins associated with vesicular trafficking at the endosome. It is recruited to the endosomes by its interaction with endofin. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOM1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TOM1
Gene Accession No. O60784, O88746
Gene Alias FLJ33404; target of myb 1; target of Myb protein 1; target of myb1; target of myb1 (chicken); target of myb1 homolog; target of myb1 homolog (chicken); target of myb1 membrane trafficking protein; TOM1
Gene Symbols TOM1
Host Species Rabbit
Immunogen Recombinant Human Target of Myb protein 1 protein (306-487AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10043, 21968
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.