Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOM1L1 Antibody (TOM1L1/4690), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308878B
Description
TOM1L1 Monoclonal antibody specifically detects TOM1L1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
TOM1L1 | |
Monoclonal | |
Biotin | |
OK/KNS-CL.3, Src-activating and signaling molecule protein, SRCASMSrc activating and signaling molecule, target of myb 1-like 1, target of myb1 (chicken) homolog-like 1, target of myb1 (chicken)-like 1, target of myb1-like 1, Target of Myb-like protein 1, TOM1-like protein 1 | |
Recombinant full-length human TOM1L1 protein (Uniprot: O75674) | |
100 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
TOM1L1/4690 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
10040 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction