Learn More
Invitrogen™ TOM1L1 Monoclonal Antibody (3F12)

Description
Positive Control: human TOM1L1-transfected 293T Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | TOM1L1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 3F12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | TOM1L1 |
| Gene Accession No. | O75674 |
| Gene Alias | 2310045L10Rik; C80573; Jerry protein; OK/KNS-CL.3; RGD1562626; Src activating and signaling molecule; src-activating and signaling molecule protein; SRCASM; target of myb1 (chicken)-like 1; target of myb1 like 1 membrane trafficking protein; target of myb1-like 1; target of myb1-like 1 (chicken); Target of Myb-like protein 1; TOM1L1; TOM1-like protein 1; Unknown (protein for MGC:142395) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.