Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOM1L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570545
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570545 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570545 Supplier Invitrogen™ Supplier No. PA570545
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

TOM1L1 is an adapter protein involved in signaling pathways. Interacts with the SH2 and SH3 domains of various signaling proteins when it is phosphorylated. May promotes FYN activation, possibly by disrupting intramolecular SH3-dependent interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOM1L1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TOM1L1
Gene Accession No. Q923U0
Gene Alias 2310045L10Rik; C80573; Jerry protein; OK/KNS-CL.3; RGD1562626; Src activating and signaling molecule; src-activating and signaling molecule protein; SRCASM; target of myb1 (chicken)-like 1; target of myb1 like 1 membrane trafficking protein; target of myb1-like 1; target of myb1-like 1 (chicken); Target of Myb-like protein 1; TOM1L1; TOM1-like protein 1; Unknown (protein for MGC:142395)
Gene Symbols TOM1L1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of mouse TOM1L1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 71943
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.