Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOM1L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554753
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554753 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554753 Supplier Invitrogen™ Supplier No. PA554753
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLHVNDDLNN VFLRYERFER YRSGRSVQNA SNGVLNEVTE DNLIDLGPGS PAVVSPMVGN TAPPSSLSSQ LAGLDLGTES VSGTLSSLQQ CNPRDGF Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

This gene belongs to a small gene family whose members have an N-terminal VHS domain followed by a GAT domain; domains which typically participate in vesicular trafficking. The canonical protein encoded by this gene also has a C-terminal clathrin binding motif. This protein has been shown to interact with Tollip, clathrin and ubiquitin and is thought to play a role in endosomal sorting. This gene resides in the 3.7 Mb deletion of chromosome region 17p11.2 that is associated with Smith-Magenis syndrome. Alternative splicing results in multiple transcript variants encoding distinct proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOM1L2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TOM1L2
Gene Accession No. Q6ZVM7
Gene Alias 2900016I08Rik; A730055F12Rik; AU042072; FLJ32746; myb1-like protein 2; Srebf1; target of myb1 like 2 membrane trafficking protein; target of myb1-like 2; target of myb1-like 2 (chicken); Target of Myb-like protein 2; Tom1l2; TOM1-like protein 2; UNQ3124
Gene Symbols TOM1L2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TOM1L2. Recombinant protein control fragment (Product #RP-93433).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146691
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.