Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOM20 Monoclonal Antibody (1D6F5), CoraLite594, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc CL59466777100UL
This item is not returnable.
View return policy
Description
TOM20 Monoclonal antibody specifically detects TOM20 in Human samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
| TOM20 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P04792 | |
| Mouse | |
| 100 μL | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles, store in dark | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry | |
| 1D6F5 | |
| CoraLite 594 | |
| HSPB1 | |
| 1810060K07Rik; BB284719; Gm19268; KIAA0016; MAS20; mitochondrial 20 kDa outer membrane protein; Mitochondrial import receptor subunit TOM20 homolog; mKIAA0016; MOM19; outer mitochondrial membrane receptor rTOM20; outer mitochondrial membrane receptor Tom20; TOM20; TOMM20; translocase of outer mitochondrial membrane 20; translocase of outer mitochondrial membrane 20 homolog (yeast); translocase of outer mitochondrial membrane 20 homolog type II | |
| Protein A | |
| RUO | |
| 3315 | |
| Liquid | |
| Human |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction