Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOM20 Polyclonal Antibody, CoraLite555, Proteintech
SDP

Catalog No. 502332705 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-233-2705 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-233-2705 Supplier Proteintech Group Inc Supplier No. CL55511802100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TOM20 Polyclonal antibody specifically detects TOM20 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry

Specifications

Antigen TOM20
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate CoraLite 555
Formulation PBS with 0.5% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Accession No. Q15388
Gene Alias 1810060K07Rik; BB284719; Gm19268; KIAA0016; MAS20; mKIAA0016; MOM19; TOM20; TOMM20
Host Species Rabbit
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9804
Target Species Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.