Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOMM20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578300
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578300 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578300 Supplier Invitrogen™ Supplier No. PA578300
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: 293T, A431, HeLa, HepG2 Predicted Reactivity: Rhesus Monkey (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The mitochondrial preprotein translocases of the outer membrane (Tom) is a multisubunit protein complex that facilitates the import of nucleus-encoded precursor proteins across the mitochondrial outer membrane. The Tom machinery consists of import receptors for the initial binding of cytosolically synthesized preproteins and a general import pore (GIP) for the membrane translocation of various preproteins into the mitochondria. The import receptors include Tom20 and Tom22, which form a heteromeric receptor complex that initiates the insertion of newly synthesized proteins into the outer membrane and then directs the precursor protein into the GIP. In yeast, Tom22 is the essential component of the import receptor complex as it functions as both a receptor for the preproteins and serves as a docking point for both Tom20 and the GIP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOMM20
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene TOMM20
Gene Accession No. Q15388, Q62760, Q9DCC8
Gene Alias 1810060K07Rik; BB284719; Gm19268; KIAA0016; MAS20; mitochondrial 20 kDa outer membrane protein; Mitochondrial import receptor subunit TOM20 homolog; mKIAA0016; MOM19; outer mitochondrial membrane receptor rTOM20; outer mitochondrial membrane receptor Tom20; TOM20; TOMM20; translocase of outer mitochondrial membrane 20; translocase of outer mitochondrial membrane 20 homolog (yeast); translocase of outer mitochondrial membrane 20 homolog type II
Gene Symbols TOMM20
Host Species Rabbit
Immunogen Full length Human TOMM20 recombinant protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 266601, 67952, 9804
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.