Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOMM20L Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | TOMM20L |
---|---|
Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TOMM20L Polyclonal antibody specifically detects TOMM20L in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
TOMM20L | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
translocase of outer mitochondrial membrane 20 homolog (yeast)-like, translocase of outer mitochondrial membrane 20 homolog type I, UNQ9438, UNQ9438/PRO34772 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: AFKRRLRDKRRAEPQKAEEQGTQLWDPTKNKKLQELFLQEVRMGELWLSRGEH | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
Polyclonal | |
Purified | |
RUO | |
PBS, pH 7.2, 40% glycerol | |
387990 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title