Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOMM22 Antibody (7C11), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP315219
Description
TOMM22 Monoclonal antibody specifically detects TOMM22 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
TOMM22 | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.4), 150mM NaCl, and 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
7C11 | |
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
1C9-2, hTom22, mitochondrial import receptor subunit TOM22 homolog, mitochondrial import receptor Tom22, MST065, TOM22MSTP065, Translocase of outer membrane 22 kDa subunit homolog, translocase of outer mitochondrial membrane 22 homolog (yeast) | |
A synthesized peptide derived from human TOMM22. UniProt ID (Q9NS69). | |
100 μg | |
Membrane Trafficking and Chaperones | |
56993 | |
Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction