Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOMM22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551804
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551804 Supplier Invitrogen™ Supplier No. PA551804
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAAAVAAAGA GEPQSPDELL PKGDAEKPEE ELEEDDDEEL DETLSERLWG LTEMFPERVR SAAGATFDLS LFVAQK Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 89%.

Tom22 has a negatively charged N-terminal region exposed to the cytosol, a putative transmembrane region, and a C-terminal intermembrane space region with little negative charge. The following functions can be assigned to the three domains of Tom22: the cytosolic N-terminal domain plays a dual role, specifically important for presequence binding; the intermembrane space domain provides a trans binding site for presequences, and the single membrane anchor of Tom22 is crucial for the integrity of the GIP (general import gene) complex. The TOM complex of mammalian mitochondria resembles the fungal Tom complex, but is distinct from the plant TOM system. Thus, while unique components of the mammalian mitochondrial import system have been identified (e. g. TOM34 and metaxin), Tom22, and Tom37 have not been identified in plant mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOMM22
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tomm22
Gene Accession No. Q9NS69
Gene Alias 1C9-2; 2310047D01; hTom22; Mitochondrial import receptor subunit TOM22 homolog; mitochondrial import receptor Tom22; MST065; MSTP065; rTOM22; Tom22; Tomm22; Translocase of outer membrane 22 kDa subunit homolog; translocase of outer mitochondrial membrane 22; translocase of outer mitochondrial membrane 22 homolog; translocase of outer mitochondrial membrane 22 homolog (yeast)
Gene Symbols Tomm22
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TOMM22. Recombinant protein control fragment (Product #RP-91498).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56993
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.