Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOMM34 Antibody (OTI2A9), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274550F
Description
TOMM34 Monoclonal specifically detects TOMM34 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
TOMM34 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
TOMM34 | |
Full length recombinant protein of human TOMM34 (NP_006800) produced in HEK293 cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2A9 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
hTom34, HTOM34P, mitochondrial import receptor subunit TOM34, outer mitochondrial membrane translocase (34kD), TOM34, Translocase of outer membrane 34 kDa subunit, translocase of outer mitochondrial membrane 34, URCC3 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
10953.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction