Learn More
TOMM34 Mouse anti-Human, Clone: OTI1B4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503183

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is involved in the import of precursor proteins into mitochondria. The encoded protein has a chaperone-like activity, binding the mature portion of unfolded proteins and aiding their import into mitochondria. This protein, which is found in the cytoplasm and sometimes associated with the outer mitochondrial membrane, has a weak ATPase activity and contains 6 TPR repeats.Specifications
TOMM34 | |
Monoclonal | |
Unconjugated | |
TOMM34 | |
HTOM34P, TOM34, URCC3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10953 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1B4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q15785 | |
TOMM34 | |
Full length human recombit protein of human TOMM34 produced in HEK293 cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.