Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOMM34 Mouse anti-Human, Clone: OTI1G7, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA503216

Description
The protein encoded by this gene is involved in the import of precursor proteins into mitochondria. The encoded protein has a chaperone-like activity, binding the mature portion of unfolded proteins and aiding their import into mitochondria. This protein, which is found in the cytoplasm and sometimes associated with the outer mitochondrial membrane, has a weak ATPase activity and contains 6 TPR repeats.Specifications
TOMM34 | |
Monoclonal | |
1.2 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q15785 | |
TOMM34 | |
Full length human recombit protein of human TOMM34 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1G7 | |
Unconjugated | |
TOMM34 | |
HTOM34P, TOM34, URCC3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10953 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction