Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOMM34 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553862
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553862 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553862 Supplier Invitrogen™ Supplier No. PA553862
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LCYLVLKQYT EAVKDCTEAL KLDGKNVKAF YRRAQAHKAL KDYKSSFADI SNLLQIEPRN GPAQKLRQEV KQNL Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.

The protein encoded by this gene is involved in the import of precursor proteins into mitochondria. The encoded protein has a chaperone-like activity, binding the mature portion of unfolded proteins and aiding their import into mitochondria. This protein, which is found in the cytoplasm and sometimes associated with the outer mitochondrial membrane, has a weak ATPase activity and contains 6 TPR repeats.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOMM34
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tomm34
Gene Accession No. Q15785
Gene Alias 2610100K07Rik; hTom34; HTOM34P; mitochondrial import receptor subunit TOM34; outer mitochondrial membrane translocase (34kD); TOM34; Tomm34; Translocase of outer membrane 34 kDa subunit; translocase of outer mitochondrial membrane 34; URCC3
Gene Symbols Tomm34
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TOMM34. Recombinant protein control fragment (Product #RP-92447).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10953
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.