Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOMM40 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519245
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519245 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519245 Supplier Invitrogen™ Supplier No. PA519245
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

TOMM40 is the channel-forming subunit of the translocase of the mitochondrial outer membrane complex that is essential for protein import into mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOMM40
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene TOMM40
Gene Accession No. O96008
Gene Alias AW539759; C19orf1; D19S1177E; haymaker protein; mitochondrial import receptor subunit TOM40 homolog; mitochondrial outer membrane protein; mitochondrial outer membrane protein 35; mitochondrial outer membrane protein of 35 kDa; Mitochondrial outer membrane protein of 38 kDa; mitochondrial translocase of outer membrane protein; Mom35; OM38; outer membrane protein 38 kDa; p38.5; PER EC1; PEREC1; PER-EC1; Protein Haymaker; TOM40; Tom40a; Tomm40; Translocase of outer membrane 40 kDa subunit homolog; translocase of outer mitochondrial membrane 40; translocase of outer mitochondrial membrane 40 homolog; translocase of outer mitochondrial membrane 40 homolog (yeast)
Gene Symbols TOMM40
Host Species Goat
Immunogen Synthetic peptide sequence (HTVALSTIGESNYH) corresponding to the internal amino acids of TOMM40 (aa 117-131).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10452
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.