Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOMM40 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578325
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578325 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578325 Supplier Invitrogen™ Supplier No. PA578325
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: 293T, A431, HeLa, HepG2 Predicted Reactivity: Pig (100%), Rhesus Monkey (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TOMM40 is the channel-forming subunit of the translocase of the mitochondrial outer membrane complex that is essential for protein import into mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOMM40
Applications Western Blot
Classification Polyclonal
Concentration 0.94 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene TOMM40
Gene Accession No. O96008
Gene Alias AW539759; C19orf1; D19S1177E; haymaker protein; mitochondrial import receptor subunit TOM40 homolog; mitochondrial outer membrane protein; mitochondrial outer membrane protein 35; mitochondrial outer membrane protein of 35 kDa; Mitochondrial outer membrane protein of 38 kDa; mitochondrial translocase of outer membrane protein; Mom35; OM38; outer membrane protein 38 kDa; p38.5; PER EC1; PEREC1; PER-EC1; Protein Haymaker; TOM40; Tom40a; Tomm40; Translocase of outer membrane 40 kDa subunit homolog; translocase of outer mitochondrial membrane 40; translocase of outer mitochondrial membrane 40 homolog; translocase of outer mitochondrial membrane 40 homolog (yeast)
Gene Symbols TOMM40
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Human TOMM40. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10452
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.